Assay Details
| Gene Symbol: BGLU15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337086.1 | NP_001325181.1 | 6 - 7 | 960 | 104 | |
| NM_130008.3 | NP_181973.1 | 11 - 12 | 1709 | 104 | ||
More Information
Set Membership: |
|

BGLU15
Genomic Map