Assay Details
| Gene Symbol: PEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130012.4 | NP_181977.1 | 5 - 6 | 687 | 74 | |
| GenBank mRNA | AY091016.1 | - | 5 - 6 | 556 | 74 | |
| BT000990.1 | - | 4 - 5 | 467 | 74 | ||
More Information
Set Membership: |
|

PEN2
Genomic Map