Assay Details
| Gene Symbol: PEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130012.4 | NP_181977.1 | 6 - 7 | 943 | 81 | |
| GenBank mRNA | AY091016.1 | - | 6 - 7 | 812 | 81 | |
| BT000990.1 | - | 5 - 6 | 723 | 81 | ||
More Information
Set Membership: |
|

PEN2
Genomic Map