Assay Details
Gene Symbol: HB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130055.2 | NP_182018.1 | 1 - 2 | 411 | 87 | |
GenBank mRNA | AB493589.1 | - | 1 - 2 | 205 | 87 | |
AJ441251.1 | - | 1 - 2 | 205 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
