Assay Details
| Gene Symbol: HB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130055.2 | NP_182018.1 | 2 - 3 | 768 | 103 | |
| GenBank mRNA | AB493589.1 | - | 2 - 3 | 562 | 103 | |
| AJ441251.1 | - | 2 - 3 | 562 | 103 | ||
More Information
Set Membership: |
|

HB4
Genomic Map