Assay Details
| Gene Symbol: GAE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130094.2 | NP_182056.1 | 1 | 839 | 90 | |
| GenBank mRNA | AY594693.1 | - | 1 | 268 | 90 | |
| BT004225.1 | - | 1 | 657 | 90 | ||
| BT005652.1 | - | 1 | 268 | 90 | ||
More Information
Set Membership: |
|

GAE4
Genomic Map