Assay Details
| Gene Symbol: BEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337127.1 | NP_001323894.1 | 2 - 3 | 923 | 100 | |
| NM_001337128.1 | NP_001323895.1 | 1 - 2 | 587 | 100 | ||
| NM_130102.5 | NP_182064.1 | 2 - 3 | 923 | 100 | ||
More Information
Set Membership: |
|

BEN1
Genomic Map