Assay Details
| Gene Symbol: ROPGEF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130152.3 | NP_182113.2 | 3 - 4 | 697 | 158 | |
| GenBank mRNA | AK229390.1 | - | 3 - 4 | 601 | 158 | |
More Information
Set Membership: |
|

ROPGEF4
Genomic Map