Assay Details
| Gene Symbol: PIP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084597.1 | NP_001078066.1 | 2 - 3 | 777 | 79 | |
| NM_001084598.1 | NP_001078067.1 | 2 - 3 | 777 | 79 | ||
| NM_130159.4 | NP_182120.1 | 2 - 3 | 991 | 79 | ||
| GenBank mRNA | AK316878.1 | - | 2 - 3 | 681 | 79 | |
| AY048294.1 | - | 2 - 3 | 682 | 79 | ||
| AY063931.1 | - | 2 - 3 | 682 | 79 | ||
| AY091254.1 | - | 2 - 3 | 622 | 79 | ||
| HM217349.1 | - | 2 - 3 | 659 | 79 | ||
| X68293.1 | - | 676 | 79 | |||
More Information
Set Membership: |
|

PIP1B
Genomic Map