Assay Details
| Gene Symbol: BRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337166.1 | NP_001318432.1 | 5 - 6 | 3433 | 101 | |
| NM_001337167.1 | NP_001323762.1 | 5 - 6 | 3562 | 101 | ||
| NM_001337168.1 | NP_001323760.1 | 5 - 6 | 3407 | 101 | ||
| NM_001337169.1 | NP_001323761.1 | 5 - 6 | 3536 | 101 | ||
| NM_130165.3 | NP_182126.2 | 4 - 5 | 2867 | 101 | ||
| NM_201966.2 | NP_973695.1 | 4 - 5 | 2867 | 101 | ||
| GenBank mRNA | AJ703891.1 | - | 4 - 5 | 2867 | 101 | |
More Information
Set Membership: |
|

BRM
Genomic Map