Assay Details
| Gene Symbol: BRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337166.1 | NP_001318432.1 | 10 - 11 | 3985 | 80 | |
| NM_001337167.1 | NP_001323762.1 | 10 - 11 | 4117 | 80 | ||
| NM_001337168.1 | NP_001323760.1 | 10 - 11 | 3962 | 80 | ||
| NM_001337169.1 | NP_001323761.1 | 10 - 11 | 4088 | 80 | ||
| NM_130165.3 | NP_182126.2 | 9 - 10 | 3419 | 80 | ||
| NM_201966.2 | NP_973695.1 | 9 - 10 | 3422 | 80 | ||
| GenBank mRNA | AJ703891.1 | - | 9 - 10 | 3422 | 80 | |
More Information
Set Membership: |
|

BRM
Genomic Map