Assay Details
| Gene Symbol: UBC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337170.1 | NP_001318433.1 | 5 - 6 | 781 | 84 | |
| NM_001337171.1 | NP_001324870.1 | 4 - 5 | 911 | 84 | ||
| NM_001337172.1 | NP_001324871.1 | 5 - 6 | 654 | 84 | ||
| NM_001337173.1 | NP_001324869.1 | 3 - 4 | 540 | 84 | ||
| NM_130166.4 | NP_566062.1 | 4 - 5 | 410 | 84 | ||
| GenBank mRNA | AK228245.1 | - | 4 - 5 | 365 | 84 | |
| BT005208.1 | - | 4 - 5 | 287 | 84 | ||
| DQ027021.1 | - | 4 - 5 | 287 | 84 | ||
More Information
Set Membership: |
|

UBC6
Genomic Map