Assay Details
| Gene Symbol: MPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202826.1 | NP_001189755.1 | 3 - 4 | 485 | 72 | |
| NM_001337177.1 | NP_001325006.1 | 3 - 4 | 729 | 72 | ||
| NM_130170.4 | NP_182131.2 | 3 - 4 | 485 | 72 | ||
| GenBank mRNA | AK117449.1 | - | 3 - 4 | 474 | 72 | |
| BT024898.1 | - | 3 - 4 | 432 | 72 | ||
More Information
Set Membership: |
|

MPK12
Genomic Map