Assay Details
Gene Symbol: ABIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125062.1 | NP_001118534.1 | 2 - 3 | 370 | 99 | |
NM_001202827.1 | NP_001189756.1 | 2 - 3 | 377 | 99 | ||
NM_130185.3 | NP_566067.1 | 2 - 3 | 370 | 99 | ||
GenBank mRNA | AY085146.1 | - | 2 - 3 | 342 | 99 | |
AY817012.1 | - | 2 - 3 | 175 | 99 | ||
BT025863.1 | - | 2 - 3 | 175 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
