Assay Details
Gene Symbol: SPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337186.1 | NP_001325255.1 | 3 - 4 | 2165 | 82 | |
NM_130197.5 | NP_182157.2 | 3 - 4 | 1986 | 82 | ||
GenBank mRNA | AF135455.1 | - | 3 - 4 | 1759 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
