Assay Details
| Gene Symbol: HB-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130233.5 | NP_182191.1 | 1 - 2 | 558 | 84 | |
| GenBank mRNA | AY045826.1 | - | 1 - 2 | 540 | 84 | |
| AY091364.1 | - | 1 - 2 | 364 | 84 | ||
More Information
Set Membership: |
|

HB-7
Genomic Map