Assay Details
Gene Symbol: NF-YC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331746.1 | NP_001320567.1 | 1 - 2 | 522 | 66 | |
NM_100672.3 | NP_563803.1 | 1 - 2 | 637 | 66 | ||
GenBank mRNA | AY086815.1 | - | 1 - 2 | 436 | 66 | |
BT025266.1 | - | 1 - 2 | 419 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
