Assay Details
Gene Symbol: NF-YC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100672.3 | NP_563803.1 | 2 - 3 | 721 | 103 | |
GenBank mRNA | AY086815.1 | - | 2 - 3 | 520 | 103 | |
BT025266.1 | - | 2 - 3 | 503 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
