Assay Details
Gene Symbol: NF-YC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331746.1 | NP_001320567.1 | 2 - 3 | 887 | 65 | |
NM_100672.3 | NP_563803.1 | 3 - 4 | 765 | 65 | ||
GenBank mRNA | AY086815.1 | - | 3 - 4 | 564 | 65 | |
BT025266.1 | - | 3 - 4 | 547 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
