Assay Details
Gene Symbol: ACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100683.3 | NP_172287.1 | 1 - 2 | 170 | 75 | |
GenBank mRNA | AY122907.1 | - | 1 - 2 | 105 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
