Assay Details
| Gene Symbol: BSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198006.1 | NP_001184935.1 | 10 - 11 | 1719 | 88 | |
| NM_100715.4 | NP_172318.1 | 10 - 11 | 1812 | 88 | ||
More Information
Set Membership: |
|

BSL2
Genomic Map