Assay Details
Gene Symbol: CRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198007.1 | NP_001184936.1 | 3 - 4 | 506 | 72 | |
NM_100718.5 | NP_563816.1 | 3 - 4 | 571 | 72 | ||
GenBank mRNA | AY056320.1 | - | 3 - 4 | 439 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
