Assay Details
Gene Symbol: SNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100773.4 | NP_563834.1 | 9 - 10 | 1764 | 71 | |
GenBank mRNA | AF439826.1 | - | 9 - 10 | 1717 | 71 | |
BT000625.1 | - | 9 - 10 | 1114 | 71 | ||
DQ132632.1 | - | 9 - 10 | 1717 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
