Assay Details
| Gene Symbol: CRT1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100791.4 | NP_172392.1 | 4 - 5 | 1008 | 91 | |
| GenBank mRNA | AY045656.1 | - | 4 - 5 | 703 | 91 | |
| AY059662.1 | - | 4 - 5 | 661 | 91 | ||
More Information
Set Membership: |
|

CRT1b
Genomic Map