Assay Details
| Gene Symbol: CRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100804.4 | NP_172405.1 | 1 - 2 | 337 | 71 | |
| GenBank mRNA | AF325043.2 | - | 1 - 2 | 109 | 71 | |
| AF428282.1 | - | 1 - 2 | 122 | 71 | ||
| AY035050.1 | - | 1 - 2 | 179 | 71 | ||
| AY070022.1 | - | 1 - 2 | 105 | 71 | ||
More Information
Set Membership: |
|

CRB
Genomic Map