Assay Details
| Gene Symbol: CRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331830.1 | NP_001322308.1 | 6 - 7 | 1098 | 108 | |
| NM_100804.4 | NP_172405.1 | 6 - 7 | 758 | 108 | ||
| GenBank mRNA | AF325043.2 | - | 6 - 7 | 530 | 108 | |
| AF428282.1 | - | 6 - 7 | 543 | 108 | ||
| AY035050.1 | - | 6 - 7 | 600 | 108 | ||
| AY070022.1 | - | 6 - 7 | 526 | 108 | ||
More Information
Set Membership: |
|

CRB
Genomic Map