Assay Details
| Gene Symbol: CRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331830.1 | NP_001322308.1 | 7 - 8 | 1331 | 83 | |
| NM_100804.4 | NP_172405.1 | 7 - 8 | 991 | 83 | ||
| GenBank mRNA | AF325043.2 | - | 7 - 8 | 763 | 83 | |
| AF428282.1 | - | 7 - 8 | 776 | 83 | ||
| AY035050.1 | - | 7 - 8 | 833 | 83 | ||
| AY070022.1 | - | 7 - 8 | 759 | 83 | ||
More Information
Set Membership: |
|

CRB
Genomic Map