Assay Details
| Gene Symbol: CRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331830.1 | NP_001322308.1 | 9 - 10 | 1534 | 66 | |
| NM_100804.4 | NP_172405.1 | 9 - 10 | 1194 | 66 | ||
| GenBank mRNA | AF325043.2 | - | 9 - 10 | 966 | 66 | |
| AF428282.1 | - | 9 - 10 | 979 | 66 | ||
| AY035050.1 | - | 9 - 10 | 1036 | 66 | ||
| AY070022.1 | - | 9 - 10 | 962 | 66 | ||
More Information
Set Membership: |
|

CRB
Genomic Map