Assay Details
| Gene Symbol: G6PD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100813.2 | NP_563844.1 | 9 - 10 | 1337 | 76 | |
| GenBank mRNA | AK229085.1 | - | 9 - 10 | 1354 | 76 | |
| AY056232.1 | - | 9 - 10 | 1322 | 76 | ||
| AY117271.1 | - | 9 - 10 | 1233 | 76 | ||
More Information
Set Membership: |
|

G6PD4
Genomic Map