Assay Details
| Gene Symbol: BCAT-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035938.2 | NP_001031015.1 | 3 - 4 | 220 | 87 | |
| NM_001035939.3 | NP_001031016.2 | 2 - 3 | 330 | 87 | ||
| NM_100881.3 | NP_172478.1 | 2 - 3 | 304 | 87 | ||
| GenBank mRNA | AF370135.1 | - | 2 - 3 | 301 | 87 | |
| AJ271731.1 | - | 2 - 3 | 188 | 87 | ||
| AK317528.1 | - | 3 - 4 | 221 | 87 | ||
| AK319062.1 | - | 2 - 3 | 327 | 87 | ||
| AY051038.1 | - | 2 - 3 | 136 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001035939.3 transcripts.
Set Membership: |
|

BCAT-2
Genomic Map