Assay Details
| Gene Symbol: BCAT-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035938.2 | NP_001031015.1 | 5 - 6 | 579 | 84 | |
| NM_001035939.3 | NP_001031016.2 | 4 - 5 | 689 | 84 | ||
| NM_100881.3 | NP_172478.1 | 4 - 5 | 663 | 84 | ||
| GenBank mRNA | AF370135.1 | - | 4 - 5 | 660 | 84 | |
| AJ271731.1 | - | 4 - 5 | 547 | 84 | ||
| AK317528.1 | - | 5 - 6 | 580 | 84 | ||
| AK319062.1 | - | 4 - 5 | 686 | 84 | ||
| AY051038.1 | - | 4 - 5 | 495 | 84 | ||
More Information
Set Membership: |
|

BCAT-2
Genomic Map