Assay Details
| Gene Symbol: BCAT-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035938.2 | NP_001031015.1 | 8 - 9 | 926 | 69 | |
| NM_001035939.3 | NP_001031016.2 | 7 - 8 | 1036 | 69 | ||
| NM_100881.3 | NP_172478.1 | 7 - 8 | 1010 | 69 | ||
| GenBank mRNA | AF370135.1 | - | 7 - 8 | 1007 | 69 | |
| AJ271731.1 | - | 7 - 8 | 894 | 69 | ||
| AK317528.1 | - | 8 - 9 | 927 | 69 | ||
| AK319062.1 | - | 7 - 8 | 1033 | 69 | ||
| AY051038.1 | - | 7 - 8 | 842 | 69 | ||
More Information
Set Membership: |
|

BCAT-2
Genomic Map