Assay Details
| Gene Symbol: FRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331895.1 | NP_001321860.1 | 2 - 3 | 2007 | 73 | |
| NM_100898.3 | NP_563865.1 | 3 - 4 | 1537 | 73 | ||
| GenBank mRNA | AF424595.1 | - | 3 - 4 | 1522 | 73 | |
| BT000486.1 | - | 2 - 3 | 1425 | 73 | ||
More Information
Set Membership: |
|

FRS11
Genomic Map