Assay Details
| Gene Symbol: emb2004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100925.2 | NP_563871.1 | 11 - 12 | 1362 | 100 | |
| GenBank mRNA | AY056236.1 | - | 11 - 12 | 1358 | 100 | |
| AY091215.1 | - | 11 - 12 | 1226 | 100 | ||
More Information
Set Membership: |
|

emb2004
Genomic Map