Assay Details
| Gene Symbol: emb2004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100925.2 | NP_563871.1 | 5 - 6 | 861 | 149 | |
| GenBank mRNA | AY056236.1 | - | 5 - 6 | 857 | 149 | |
| AY091215.1 | - | 5 - 6 | 725 | 149 | ||
More Information
Set Membership: |
|

emb2004
Genomic Map