Assay Details
Gene Symbol: PRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331922.1 | NP_001320728.1 | 2 - 3 | 764 | 118 | |
NM_100946.3 | NP_172540.1 | 2 - 3 | 764 | 118 | ||
GenBank mRNA | AY058091.1 | - | 2 - 3 | 753 | 118 | |
AY090310.1 | - | 2 - 3 | 711 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
