Assay Details
Gene Symbol: PRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331922.1 | NP_001320728.1 | 5 - 6 | 1199 | 67 | |
NM_100946.3 | NP_172540.1 | 5 - 6 | 1051 | 67 | ||
GenBank mRNA | AY058091.1 | - | 5 - 6 | 1040 | 67 | |
AY090310.1 | - | 5 - 6 | 998 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
