Assay Details
| Gene Symbol: PRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331922.1 | NP_001320728.1 | 6 - 7 | 1307 | 72 | |
| NM_100946.3 | NP_172540.1 | 6 - 7 | 1159 | 72 | ||
| GenBank mRNA | AY058091.1 | - | 6 - 7 | 1148 | 72 | |
| AY090310.1 | - | 6 - 7 | 1106 | 72 | ||
More Information
Set Membership: |
|

PRS3
Genomic Map