Assay Details
| Gene Symbol: RECQ4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100968.3 | NP_172562.2 | 24 - 25 | 3459 | 72 | |
| GenBank mRNA | AY120761.1 | - | 24 - 25 | 3357 | 72 | |
| BT010133.1 | - | 23 - 24 | 3153 | 72 | ||
More Information
Set Membership: |
|

RECQ4A
Genomic Map