Assay Details
| Gene Symbol: AT1G11060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100979.4 | NP_172573.2 | 8 - 9 | 2122 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G11060| Gene Symbol: AT1G11060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100979.4 | NP_172573.2 | 8 - 9 | 2122 | 123 | |
Set Membership: |
|