Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198029.1 | NP_001184958.1 | 10 - 11 | 3495 | 80 | |
| NM_001331954.1 | NP_001320182.1 | 10 - 11 | 3810 | 80 | ||
| NM_001331955.1 | NP_001320184.1 | 10 - 11 | 3782 | 80 | ||
| NM_001331956.1 | NP_001320183.1 | 9 - 10 | 2947 | 80 | ||
| NM_100983.5 | NP_172577.2 | 11 - 12 | 3424 | 80 | ||
| GenBank mRNA | AB493447.1 | - | 10 - 11 | 3498 | 80 | |
More Information
Set Membership: |
|

AT1G11100
Genomic Map