Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198029.1 | NP_001184958.1 | 8 - 9 | 2685 | 95 | |
| NM_001331954.1 | NP_001320182.1 | 8 - 9 | 3000 | 95 | ||
| NM_001331955.1 | NP_001320184.1 | 8 - 9 | 2972 | 95 | ||
| NM_001331956.1 | NP_001320183.1 | 7 - 8 | 2137 | 95 | ||
| NM_100983.5 | NP_172577.2 | 9 - 10 | 2614 | 95 | ||
| GenBank mRNA | AB493447.1 | - | 8 - 9 | 2688 | 95 | |
More Information
Set Membership: |
|

AT1G11100
Genomic Map