Assay Details
Gene Symbol: SUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331960.1 | NP_001321706.1 | 12 - 13 | 1918 | 70 | |
NM_100986.4 | NP_172580.2 | 13 - 14 | 2402 | 70 | ||
GenBank mRNA | AF399923.1 | - | 13 - 14 | 2191 | 70 | |
AK227100.1 | - | 13 - 14 | 2203 | 70 | ||
AY091019.1 | - | 13 - 14 | 2253 | 70 | ||
AY117318.1 | - | 13 - 14 | 1975 | 70 | ||
FJ708632.1 | - | 13 - 14 | 1975 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
