Assay Details
Gene Symbol: SUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100986.4 | NP_172580.2 | 1 - 2 | 519 | 75 | |
GenBank mRNA | AF399923.1 | - | 1 - 2 | 308 | 75 | |
AK227100.1 | - | 1 - 2 | 320 | 75 | ||
AY091019.1 | - | 1 - 2 | 370 | 75 | ||
AY117318.1 | - | 1 - 2 | 92 | 75 | ||
FJ708632.1 | - | 1 - 2 | 92 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
