Assay Details
Gene Symbol: SUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331960.1 | NP_001321706.1 | 3 - 4 | 301 | 68 | |
NM_100986.4 | NP_172580.2 | 4 - 5 | 785 | 68 | ||
GenBank mRNA | AF399923.1 | - | 4 - 5 | 574 | 68 | |
AK227100.1 | - | 4 - 5 | 586 | 68 | ||
AY091019.1 | - | 4 - 5 | 636 | 68 | ||
AY117318.1 | - | 4 - 5 | 358 | 68 | ||
FJ708632.1 | - | 4 - 5 | 358 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
