Assay Details
Gene Symbol: SUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331960.1 | NP_001321706.1 | 5 - 6 | 443 | 115 | |
NM_100986.4 | NP_172580.2 | 6 - 7 | 927 | 115 | ||
GenBank mRNA | AF399923.1 | - | 6 - 7 | 716 | 115 | |
AK227100.1 | - | 6 - 7 | 728 | 115 | ||
AY091019.1 | - | 6 - 7 | 778 | 115 | ||
AY117318.1 | - | 6 - 7 | 500 | 115 | ||
FJ708632.1 | - | 6 - 7 | 500 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
