Assay Details
Gene Symbol: SUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331960.1 | NP_001321706.1 | 8 - 9 | 655 | 86 | |
NM_100986.4 | NP_172580.2 | 9 - 10 | 1139 | 86 | ||
GenBank mRNA | AF399923.1 | - | 9 - 10 | 928 | 86 | |
AK227100.1 | - | 9 - 10 | 940 | 86 | ||
AY091019.1 | - | 9 - 10 | 990 | 86 | ||
AY117318.1 | - | 9 - 10 | 712 | 86 | ||
FJ708632.1 | - | 9 - 10 | 712 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
