Assay Details
| Gene Symbol: STP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100998.4 | NP_172592.1 | 3 - 4 | 1305 | 80 | |
| GenBank mRNA | AY054249.1 | - | 3 - 4 | 1193 | 80 | |
| AY059781.1 | - | 3 - 4 | 1211 | 80 | ||
| AY133845.1 | - | 3 - 4 | 1081 | 80 | ||
More Information
Set Membership: |
|

STP1
Genomic Map