Assay Details
| Gene Symbol: AT5G56480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125031.2 | NP_200459.1 | 1 - 2 | 303 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G56480| Gene Symbol: AT5G56480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125031.2 | NP_200459.1 | 1 - 2 | 303 | 63 | |
Set Membership: |
|