Assay Details
| Gene Symbol: PUM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345201.1 | NP_001318812.1 | 2 - 3 | 1497 | 118 | |
| NM_001345202.1 | NP_001332601.1 | 2 - 3 | 1497 | 118 | ||
| NM_001345203.1 | NP_001332600.1 | 2 - 3 | 1229 | 118 | ||
| NM_001345204.1 | NP_001332599.1 | 2 - 3 | 1229 | 118 | ||
| NM_001345205.1 | NP_001332598.1 | 1 - 2 | 1123 | 118 | ||
| NM_125034.2 | NP_200462.2 | 1 - 2 | 1102 | 118 | ||
More Information
Set Membership: |
|

PUM12
Genomic Map